Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027391.8 | NP_001022562.1 | 6 - 7 | 894 | 71 | |
NM_001027392.6 | NP_001022563.1 | 6 - 7 | 894 | 71 | ||
GenBank mRNA | AF256466.1 | - | 6 - 7 | 884 | 71 | |
AF256467.1 | - | 6 - 7 | 884 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |