Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ras-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065571.6 | NP_497972.1 | 2 - 3 | 220 | 73 | |
GenBank mRNA | U80675.1 | - | 2 - 3 | 215 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |