Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fsn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065645.7 | NP_498046.1 | 8 - 9 | 1915 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |