Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: larp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047402.3 | NP_001040867.2 | 1 - 2 | 126 | 59 | |
NM_001047403.5 | NP_001040868.3 | 3 - 4 | 894 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |