Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prmt-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065711.8 | NP_498112.1 | 6 - 7 | 1747 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |