Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065718.8 | NP_498119.1 | 7 - 8 | 1194 | 148 | |
GenBank mRNA | AY052771.1 | - | 7 - 8 | 1194 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |