Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gro-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065721.7 | NP_498122.2 | 1 - 2 | 189 | 100 | |
GenBank mRNA | AY052773.1 | - | 1 - 2 | 189 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |