Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ucr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356762.3 | NP_001343795.1 | 5 | 838 | 62 | |
NM_001379716.1 | NP_001368667.1 | 2 - 3 | 313 | 62 | ||
NM_001379717.1 | NP_001368666.1 | 2 - 3 | 313 | 62 | ||
NM_065801.8 | NP_498202.1 | 5 - 6 | 861 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |