Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129200.4 | NP_001122672.1 | 2 - 3 | 321 | 98 | |
NM_065828.8 | NP_498229.3 | 2 - 3 | 446 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |