Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ida-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065844.6 | NP_498245.2 | 1 - 2 | 166 | 111 | |
GenBank mRNA | AF126740.1 | - | 1 - 2 | 123 | 111 | |
AJ245560.1 | - | 1 - 2 | 123 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |