Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330885.2 | NP_001317785.1 | 6 - 7 | 719 | 154 | |
NM_001330886.3 | NP_001317786.1 | 4 - 5 | 443 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |