Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361719.2 | NP_001348731.1 | 10 - 11 | 1771 | 89 | |
NM_001361720.3 | NP_001348732.1 | 7 - 8 | 994 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |