Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027395.5 | NP_001022566.1 | 3 - 4 | 460 | 77 | |
NM_001027396.5 | NP_001022567.1 | 3 - 4 | 262 | 77 | ||
NM_001379772.1 | NP_001367825.1 | 5 - 6 | 973 | 77 | ||
NM_066066.7 | NP_498467.3 | 5 - 6 | 936 | 77 | ||
GenBank mRNA | AF399852.1 | - | 2 - 3 | 240 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |