Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027452.4 | NP_001022623.1 | 4 - 5 | 1004 | 73 | |
NM_001027453.4 | NP_001022624.1 | 1 - 2 | 140 | 73 | ||
GenBank mRNA | Z12116.1 | - | 1065 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |