Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027384.6 | NP_001022555.1 | 4 - 5 | 971 | 82 | |
GenBank mRNA | U01303.1 | - | 4 - 5 | 971 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |