Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066142.5 | NP_498543.1 | 2 - 3 | 259 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |