Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mls-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066239.4 | NP_498640.1 | 4 - 5 | 641 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |