Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cup-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027550.4 | NP_001022721.2 | 6 - 7 | 1400 | 91 | |
NM_001027551.5 | NP_001022722.2 | 5 - 6 | 1340 | 91 | ||
NM_001268016.4 | NP_001254945.1 | 5 - 6 | 1352 | 91 | ||
NM_001382919.1 | 5 - 6 | 1349 | 91 | |||
NM_066263.6 | NP_498664.3 | 6 - 7 | 1365 | 91 | ||
GenBank mRNA | AF482952.1 | - | 5 - 6 | 1346 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |