Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047386.5 | NP_001040851.1 | 5 - 6 | 977 | 147 | |
NM_001268026.4 | NP_001254955.1 | 5 - 6 | 988 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |