Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066427.7 | NP_498828.2 | 1 - 2 | 96 | 82 | |
GenBank mRNA | AJ539470.1 | - | 1 - 2 | 89 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |