Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066427.7 | NP_498828.2 | 2 - 3 | 202 | 87 | |
GenBank mRNA | AJ539470.1 | - | 2 - 3 | 195 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |