Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066427.7 | NP_498828.2 | 8 - 9 | 2090 | 136 | |
GenBank mRNA | AJ539470.1 | - | 8 - 9 | 2083 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |