Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066465.5 | NP_498866.1 | 1 - 2 | 188 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |