Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066500.6 | NP_498901.1 | 2 - 3 | 207 | 139 | |
NM_066501.4 | NP_498902.1 | 2 - 3 | 207 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |