Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066502.6 | NP_498903.3 | 9 - 10 | 961 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |