Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066606.6 | NP_499007.1 | 9 - 10 | 4250 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |