Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066613.5 | NP_499014.1 | 4 - 5 | 1305 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |