Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066653.4 | NP_499054.1 | 1 - 2 | 146 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |