Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025962.3 | NP_001021133.1 | 9 - 10 | 2022 | 70 | |
NM_001025963.6 | NP_001021134.1 | 10 - 11 | 2205 | 70 | ||
NM_001306716.3 | NP_001293645.1 | 6 - 7 | 1706 | 70 | ||
NM_066679.5 | NP_499080.3 | 8 - 9 | 3314 | 70 | ||
GenBank mRNA | AF241656.1 | - | 9 - 10 | 2075 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |