Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066763.7 | NP_499164.1 | 5 - 6 | 785 | 143 | |
GenBank mRNA | AJ005867.1 | - | 5 - 6 | 788 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |