Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306672.3 | NP_001293601.1 | 1 - 2 | 227 | 121 | |
NM_066775.6 | NP_499176.1 | 3 - 4 | 770 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |