Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cut-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066999.7 | NP_499400.2 | 2 - 3 | 132 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |