Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067004.7 | NP_499405.4 | 2 - 3 | 333 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |