Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379928.1 | NP_001367300.1 | 3 - 4 | 248 | 104 | |
NM_001379929.1 | NP_001367172.1 | 2 - 3 | 578 | 104 | ||
NM_001382996.1 | 3 - 4 | 325 | 104 | |||
NM_067047.6 | NP_499448.1 | 3 - 4 | 396 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |