Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379928.1 | NP_001367300.1 | 8 - 9 | 847 | 61 | |
NM_001379929.1 | NP_001367172.1 | 7 - 8 | 1177 | 61 | ||
NM_001382996.1 | 8 - 9 | 924 | 61 | |||
NM_067047.6 | NP_499448.1 | 8 - 9 | 995 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |