Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027713.5 | NP_001022884.1 | 1 - 2 | 174 | 146 | |
NM_001372661.1 | NP_001359517.1 | 1 - 2 | 140 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |