Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nft-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067155.8 | NP_499556.1 | 6 - 7 | 994 | 77 | |
GenBank mRNA | AF069986.1 | - | 6 - 7 | 992 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |