Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyg-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347293.3 | NP_001334209.1 | 1 - 2 | 633 | 65 | |
NM_067170.7 | NP_499571.2 | 3 - 4 | 1177 | 65 | ||
GenBank mRNA | AJ319870.1 | - | 2 - 3 | 1092 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |