Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: php-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067172.6 | NP_499573.1 | 3 - 4 | 848 | 75 | |
GenBank mRNA | AF172092.1 | - | 2 - 3 | 606 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |