Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pstk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067226.2 | NP_499627.1 | 1 - 2 | 563 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |