Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129255.5 | NP_001122727.1 | 3 - 4 | 862 | 105 | |
NM_067274.6 | NP_499675.1 | 3 - 4 | 858 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |