Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067339.5 | NP_499740.1 | 6 - 7 | 956 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |