Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cua-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268273.3 | NP_001255202.1 | 13 - 14 | 2653 | 90 | |
NM_001268274.3 | NP_001255203.1 | 10 - 11 | 2154 | 90 | ||
GenBank mRNA | D83665.1 | - | 12 - 13 | 2589 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |