Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Y38C1AA.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379994.1 | NP_001367798.1 | 10 - 11 | 3258 | 92 |
Gene Symbol: cutl-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028278.5 | NP_001023449.1 | 10 - 11 | 3269 | 92 | |
NM_001028279.6 | NP_001023450.1 | 8 - 9 | 1700 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |