Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068737.2 | NP_501138.1 | 2 - 3 | 101 | 100 | |
GenBank mRNA | U76402.1 | - | 2 - 3 | 101 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |