Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307363.3 | NP_001294292.1 | 5 - 6 | 505 | 93 | |
NM_001307364.3 | NP_001294293.1 | 4 - 5 | 511 | 93 | ||
NM_001307365.3 | NP_001294294.1 | 3 - 4 | 388 | 93 | ||
NM_068737.2 | NP_501138.1 | 5 - 6 | 508 | 93 | ||
GenBank mRNA | U76402.1 | - | 5 - 6 | 508 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |