Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306917.3 | NP_001293846.1 | 12 - 13 | 2386 | 72 | |
NM_001306918.3 | NP_001293847.1 | 11 - 12 | 2212 | 72 | ||
NM_001306919.3 | NP_001293848.1 | 10 - 11 | 2116 | 72 | ||
NM_001307143.2 | NP_001294072.1 | 9 - 10 | 2251 | 72 | ||
NM_001380315.1 | NP_001368174.1 | 9 - 10 | 2023 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |