Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ocr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068771.7 | NP_501172.2 | 8 - 9 | 1916 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |