Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nep-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028032.6 | NP_001023203.2 | 13 - 14 | 2125 | 151 | |
NM_001028034.5 | NP_001023205.2 | 11 - 12 | 1789 | 151 | ||
NM_001383127.1 | 12 - 13 | 1921 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |