Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027999.4 | NP_001023170.1 | 20 - 21 | 5298 | 77 | |
NM_001028000.5 | NP_001023171.1 | 18 - 19 | 5130 | 77 | ||
NM_001028001.5 | NP_001023172.1 | 20 - 21 | 5255 | 77 | ||
NM_001028002.4 | NP_001023173.1 | 19 - 20 | 5226 | 77 | ||
NM_001268376.4 | NP_001255305.1 | 20 - 21 | 5223 | 77 | ||
NM_001380339.1 | NP_001367466.1 | 20 - 21 | 5182 | 77 | ||
GenBank mRNA | AF168688.1 | - | 19 - 20 | 5226 | 77 | |
AJ243179.2 | - | 19 - 20 | 5248 | 77 | ||
AJ243180.2 | - | 20 - 21 | 5321 | 77 | ||
AJ243181.2 | - | 19 - 20 | 5241 | 77 | ||
AJ243182.2 | - | 20 - 21 | 5275 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |